KEY465203115 Key Organics / BIONET

3-methyl-1H-1,2,4-triazole-5-thiol

  • MDL number MFCD00179327
  • Cas Number : 7271-44-5
  • Molecular Weight : 115.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-1H-1,2,4-triazole-5-thiol
Inchi Code 1S/C3H5N3S/c1-2-4-3(7)6-5-2/h1H3,(H2,4,5,6,7)
Inchi Key OUZCWDMJTKYHCA-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104357

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-2947_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart