KEY465203177 Key Organics / BIONET

N-(2-methoxyethyl)-3-nitropyridin-2-amine

  • MDL number MFCD05670397
  • Cas Number : 866010-53-9
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-methoxyethyl)-3-nitro-2-pyridinamine
Inchi Code 1S/C8H11N3O3/c1-14-6-5-10-8-7(11(12)13)3-2-4-9-8/h2-4H,5-6H2,1H3,(H,9,10)
Inchi Key GMNGFFZTDVVXHM-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104519

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart