KEY465203287 Key Organics / BIONET

3-(2-fluorophenyl)propanoic acid

  • MDL number MFCD01310820
  • Cas Number : 1643-26-1
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104797

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-3534_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart