KEY465203317 Key Organics / BIONET

4-[(2-nitrophenyl)amino]phenol

  • MDL number MFCD00239470
  • Cas Number : 54381-08-7
  • Molecular Weight : 230.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-nitroanilino)phenol
Inchi Code 1S/C12H10N2O3/c15-10-7-5-9(6-8-10)13-11-3-1-2-4-12(11)14(16)17/h1-8,13,15H
Inchi Key HSDSBIUUVWRHTM-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104909

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-3693_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart