KEY465203324 Key Organics / BIONET

1-(diphenylmethyl)azetidine-3-carbonitrile

  • MDL number MFCD00158910
  • Cas Number : 36476-86-5
  • Molecular Weight : 248.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzhydryl-3-azetidinecarbonitrile
Inchi Code 1S/C17H16N2/c18-11-14-12-19(13-14)17(15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-10,14,17H,12-13H2
Inchi Key IXMOEAHDRKNAAG-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104922

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-3712_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart