KEY465203328 Key Organics / BIONET

3-nitro-4-propyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD05670730
  • Cas Number : 866043-56-3
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/104937

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/MS-5015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart