KEY465203337 Key Organics / BIONET

methyl 2-(benzenesulfonyl)-2-diazoacetate

  • MDL number MFCD29042760
  • Cas Number : 70972-83-7
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650340

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-00040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart