KEY465203342 Key Organics / BIONET

tert-butyl N-[(1S)-1-carbamoyl-2-methylpropyl]carbamate

  • MDL number MFCD00190841
  • Cas Number : 35150-08-4
  • Molecular Weight : 216.281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/650345

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NS-00045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart