KEY465203361 Key Organics / BIONET

3-amino-N-(3-pyridinyl)benzenecarboxamide

  • MDL number MFCD09043704
  • Cas Number : 25844-48-8
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-N-(3-pyridinyl)benzamide
Inchi Code 1S/C12H11N3O/c13-10-4-1-3-9(7-10)12(16)15-11-5-2-6-14-8-11/h1-8H,13H2,(H,15,16)
Inchi Key ROXMEGSVJDLDHD-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108303

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3186_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart