KEY465203365 Key Organics / BIONET

1-benzyl-4-azepanone

  • MDL number MFCD04038554
  • Cas Number : 1208-75-9
  • Molecular Weight : 203.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3274_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart