KEY465203388 Key Organics / BIONET

N-benzyl-N-(2-bromophenyl)amine

  • MDL number MFCD02131277
  • Cas Number : 71687-81-5
  • Molecular Weight : 262.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-benzyl-N-(2-bromophenyl)amine
Inchi Code 1S/C13H12BrN/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9,15H,10H2
Inchi Key KUSIVIJKFBJPBH-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108354

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3448_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart