KEY465203396 Key Organics / BIONET

3-chloro-1-(2-fluorophenyl)-1-propanone

  • MDL number MFCD07699515
  • Cas Number : 898767-04-9
  • Molecular Weight : 186.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-1-(2-fluorophenyl)-1-propanone
Inchi Code 1S/C9H8ClFO/c10-6-5-9(12)7-3-1-2-4-8(7)11/h1-4H,5-6H2
Inchi Key WCNSDVGUYXGZDE-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108369

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3519_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart