KEY465203400 Key Organics / BIONET

4-(4-chlorophenyl)butanoic acid

  • MDL number MFCD06810494
  • Cas Number : 4619-18-5
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-chlorophenyl)butanoic acid
Inchi Code 1S/C10H11ClO2/c11-9-6-4-8(5-7-9)2-1-3-10(12)13/h4-7H,1-3H2,(H,12,13)
Inchi Key YJMDORBRISAZSC-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108384

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3645_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart