KEY465203403 Key Organics / BIONET

(1-benzhydryl-3-azetanyl)methanamine

  • MDL number MFCD03093388
  • Cas Number : 36476-88-7
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-benzhydryl-3-azetidinyl)methanamine
Inchi Code 1S/C17H20N2/c18-11-14-12-19(13-14)17(15-7-3-1-4-8-15)16-9-5-2-6-10-16/h1-10,14,17H,11-13,18H2
Inchi Key KISVATOISQDZJU-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108388

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3680_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart