KEY465203410 Key Organics / BIONET

(2S)-2-(4-bromophenyl)-2-hydroxyacetic acid >99%ee

  • MDL number MFCD00798440
  • Cas Number : 123484-90-2
  • Molecular Weight : 231.045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/368096

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4660_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart