KEY465203413 Key Organics / BIONET

methyl (1R,8S,9R,Z)-bicyclo[6.1.0]non-4-ene-9-carboxylate

  • MDL number MFCD27937194
  • Cas Number : 61490-21-9
  • Molecular Weight : 180.247

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409265

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart