KEY465203414 Key Organics / BIONET

ethyl 4-[(3R,5S)-3,5-dimethylpiperazin-1-yl]benzoate

  • MDL number MFCD27937195
  • Cas Number : 234082-05-4
  • Molecular Weight : 262.353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/409277

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart