KEY465203415 Key Organics / BIONET

(14S,15S)-5-methoxy-15-methyltetracyclo[8.7.0.0²,7.0¹¹,¹5]heptadeca-2,4,6,12-tetraen-14-ol

  • MDL number MFCD28101540
  • Cas Number : 17980-88-0
  • Molecular Weight : 284.399

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437466

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4829_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart