KEY466131848 Key Organics / BIONET

N-(3-hydroxy-1,2,4-thiadiazol-5-yl)-4-methylbenzamide

  • MDL number MFCD04124081
  • Cas Number : 138468-80-1
  • Molecular Weight : 235.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1858

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/10L-700S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart