KEY466132394 Key Organics / BIONET

ethyl 3,5-dimethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD05974910
  • Cas Number : 103409-93-4
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/10X-0816_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart