KEY466132469 Key Organics / BIONET

(2Z)-3-[(4-fluorophenyl)amino]-2-(thiophen-3-yl)prop-2-enenitrile

  • MDL number MFCD00214693
  • Cas Number : 477851-22-2
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3820

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11B-124_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart