KEY466132470 Key Organics / BIONET

(2E)-3-[(2,4-dichlorophenyl)amino]-2-(thiophen-3-yl)prop-2-enenitrile

  • MDL number MFCD00138249
  • Cas Number : 477851-23-3
  • Molecular Weight : 295.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3825

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11B-134_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart