KEY466132479 Key Organics / BIONET

5-{4-[(2H-1,3-benzodioxol-5-yl)methyl]piperazin-1-yl}-1H-1,2,4-triazol-3-amine

  • MDL number MFCD00138993
  • Cas Number : 477851-41-5
  • Molecular Weight : 302.338

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3851

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11C-014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart