KEY466132489 Key Organics / BIONET

1-{6-[(2,4-dichlorophenyl)methyl]pyridazin-3-yl}-4-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD00140814
  • Cas Number : 321385-81-3
  • Molecular Weight : 396.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3912

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11D-054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart