KEY466132490 Key Organics / BIONET

1-(4-chlorophenyl)-3-[(1E)-(dimethylamino)methylidene]thiourea

  • MDL number MFCD00139320
  • Cas Number : 73161-60-1
  • Molecular Weight : 241.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3913

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11D-055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart