KEY466132500 Key Organics / BIONET

4-({[(2Z)-3-methoxy-5-oxo-2,5-dihydrofuran-2-ylidene]methyl}amino)benzoic acid

  • MDL number MFCD00139338
  • Cas Number : 477852-15-6
  • Molecular Weight : 261.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3955

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11D-108_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart