KEY466132501 Key Organics / BIONET

(E)-methoxy[(3-nitro-4-{[3-(trifluoromethyl)phenyl]sulfanyl}phenyl)methylidene]amine

  • MDL number MFCD00139274
  • Cas Number : 477852-19-0
  • Molecular Weight : 356.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3961

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11D-114S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart