KEY466132536 Key Organics / BIONET

6-oxo-1-{[3-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carbohydrazide

  • MDL number MFCD00140548
  • Cas Number : 477852-89-4
  • Molecular Weight : 311.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4054

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-301S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart