KEY466132542 Key Organics / BIONET

5-(5-sulfanyl-1,3,4-oxadiazol-2-yl)-1-{[3-(trifluoromethyl)phenyl]methyl}-1,2-dihydropyridin-2-one

  • MDL number MFCD00140557
  • Cas Number : 477852-99-6
  • Molecular Weight : 353.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4069

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-317S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart