KEY466132545 Key Organics / BIONET

1-[(2,4-dichlorophenyl)methyl]-N-[(methylcarbamothioyl)amino]-6-oxo-1,6-dihydropyridine-3-carboxamide

  • MDL number MFCD00140560
  • Cas Number : 477853-03-5
  • Molecular Weight : 385.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4081

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-332S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart