KEY466132548 Key Organics / BIONET

1-[(2,4-dichlorophenyl)methyl]-5-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)-1,2-dihydropyridin-2-one

  • MDL number MFCD00140564
  • Cas Number : 477853-12-6
  • Molecular Weight : 367.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4095

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-349S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart