KEY466132550 Key Organics / BIONET

4-(4-chlorophenyl)-3-methyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD00232619
  • Cas Number : 400074-90-0
  • Molecular Weight : 351.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4100

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-354S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart