KEY466132552 Key Organics / BIONET

4-(4-chlorophenyl)-1-(4-methoxyphenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD00140642
  • Cas Number : 321391-74-6
  • Molecular Weight : 313.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4102

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-357S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart