KEY466132554 Key Organics / BIONET

1-[(4-chlorophenyl)methyl]-N-[(ethylcarbamothioyl)amino]-2-oxo-1,2-dihydropyridine-3-carboxamide

  • MDL number MFCD00232622
  • Cas Number : 400074-95-5
  • Molecular Weight : 364.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-362S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart