KEY466132559 Key Organics / BIONET

1-{[3-(trifluoromethyl)phenyl]methyl}-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00141558
  • Cas Number : 79183-40-7
  • Molecular Weight : 305.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Orange to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4134

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-400S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart