KEY466132568 Key Organics / BIONET

ethyl 5-amino-3-(2-hydroxyethoxy)-1-phenyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD00172359
  • Cas Number : 321391-76-8
  • Molecular Weight : 291.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4178

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart