KEY466132569 Key Organics / BIONET

4'-phenyl-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,3,4]triazolidine]-5'-one

  • MDL number MFCD00140495
  • Cas Number : 131365-81-6
  • Molecular Weight : 279.343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4199

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-943_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart