KEY466132576 Key Organics / BIONET

ethyl N-{2-cyano-2-[(1E)-(methoxyimino)methyl]acetyl}carbamate

  • MDL number MFCD00172920
  • Cas Number : 477853-61-5
  • Molecular Weight : 213.193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-955_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart