KEY466132580 Key Organics / BIONET

5-acetyl-1-methoxy-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00172925
  • Cas Number : 477853-68-2
  • Molecular Weight : 184.151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11E-965_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart