KEY466132581 Key Organics / BIONET

methyl 5-[(1Z)-2-(4-chlorophenyl)-1-cyanoeth-1-en-1-yl]-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD00173429
  • Cas Number : 477853-70-6
  • Molecular Weight : 300.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4221

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart