KEY466132583 Key Organics / BIONET

4-(trifluoromethyl)-2-[(E)-2-{[3-(trifluoromethyl)phenyl]amino}ethenyl]-1,3-thiazole-5-carbonitrile

  • MDL number MFCD00202363
  • Cas Number : 477853-72-8
  • Molecular Weight : 363.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4224

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart