KEY466132585 Key Organics / BIONET

5-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}methyl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00141301
  • Cas Number : 303148-05-2
  • Molecular Weight : 323.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4239

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart