KEY466132586 Key Organics / BIONET

2-oxo-2-phenylethyl 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD00173204
  • Cas Number : 303148-13-2
  • Molecular Weight : 329.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4242

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-032_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart