KEY466132589 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00664252
  • Cas Number : 122890-60-2
  • Molecular Weight : 272.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4251

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart