KEY466132590 Key Organics / BIONET

(NE,E)-1-cyano-N'-{[4-(trifluoromethyl)phenyl]amino}-N-{[4-(trifluoromethyl)phenyl]imino}methanimidamide

  • MDL number MFCD00929083
  • Cas Number : 58139-96-1
  • Molecular Weight : 385.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4252

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart