KEY466132603 Key Organics / BIONET

ethyl 2-{[2-(3-chlorophenyl)-6-nitro-1H-1,3-benzodiazol-1-yl]oxy}propanoate

  • MDL number MFCD00141519
  • Cas Number : 303149-66-8
  • Molecular Weight : 389.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-427S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart