KEY466132606 Key Organics / BIONET

2-[(E)-[(4-chlorophenyl)methylidene]amino]-4-phenyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD00171743
  • Cas Number : 303149-96-4
  • Molecular Weight : 305.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4382

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-904_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart