KEY466132607 Key Organics / BIONET

11,15-dioxa-16-azatetracyclo[8.7.0.0^{2,7}.0^{13,17}]heptadeca-1(10),2(7),3,5,8-pentaene

  • MDL number MFCD01935831
  • Cas Number : 188726-76-3
  • Molecular Weight : 227.263

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4389

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-911_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart