KEY466132610 Key Organics / BIONET

ethyl 5-hydroxy-2-methyl-3-phenyl-5-(trifluoromethyl)-1,2-oxazolidine-4-carboxylate

  • MDL number MFCD00231452
  • Cas Number : 321391-90-6
  • Molecular Weight : 319.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4413

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-939_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart