KEY466132612 Key Organics / BIONET

1-(4-tert-butylphenyl)-4-chloro-6-oxo-1,6-dihydropyridazine-3-carbonitrile

  • MDL number MFCD00244217
  • Cas Number : 303150-46-1
  • Molecular Weight : 287.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/11G-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart